missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfide Nonahydrate, Honeywell Fluka™
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
3 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Color | Colorless or Yellow |
Quantity | 3 kg |
Linear Formula | Na2S·9H2O |
UN Number | 1849 |
Formula Weight | 240.18g/mol |
Percent Purity | ≥98% |
Grade | ACS Reagent |
Chemical Name or Material | Sodium Sulfide Nonahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'