missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, NISTstandard Solution 0.01N, for Volumetric Analysis, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
Packaging:
Amber glass bottle
Unit Size:
2.5 L
Description
2.5L, Amber glass bottle
Specifications
Specifications
| CAS | 10102-17-7 |
| Physical Form | Liquid |
| Quantity | 2.5 L |
| Molecular Formula | H10Na2O8S2 |
| MDL Number | 149186 |
| Synonym | sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate |
| InChI Key | PODWXQQNRWNDGD-UHFFFAOYSA-L |
| SMILES | O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
| IUPAC Name | disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
| Molecular Weight (g/mol) | 248.172 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction