Learn More
Sodium valproate, 99.9%, MP Biomedicals™
Product Code. 11484361
Click to view available options
Quantity:
10 g
25 g
100 g
Chemical Identifiers
CAS 1069-66-5
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.196
InChI Key AEQFSUDEHCCHBT-UHFFFAOYSA-M
Synonym sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin
PubChem CID 16760703
ChEBI CHEBI:9925
IUPAC Name sodium;2-propylpentanoate
SMILES CCCC(CCC)C(=O)[O-].[Na+]
Specifications
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 0.999
Formula Weight 166.2
Chemical Name or Material Sodium valproate