Learn More
Memmert™ Software Conforming to FDA AtmoCONTROL
Product Code. 17150195 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
ICO105
ICO150
ICO240
ICO50
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17150195
Brand: Memmert™ IPP260ecoFDAQ1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
  • Base licence for reading the data logging of one unit (only for units with TwinDISPLAY)
  • Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
For Use With (Application) Only for Units with TwinDISPLAY
For Use With (Equipment) IPP260eco