Learn More
Memmert™ Software Conforming to FDA AtmoCONTROL
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
ICO105
ICO150
ICO240
ICO50
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Description
- Base licence for reading the data logging of one unit (only for units with TwinDISPLAY)
- Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Specifications
| For Use With (Application) | Only for Units with TwinDISPLAY |
| For Use With (Equipment) | IPP1400eco |