missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX4 (Master Regulator of Epithelial-Mesenchymal Transition) Mouse, Clone: SOX4/2540, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 6659-MSM1-P1
This item is not returnable.
View return policy
Specifications
SOX4 (Master Regulator of Epithelial-Mesenchymal Transition) | |
Nuclear | |
SOX4/2540 | |
Unconjugated | |
Mouse | |
47kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
6659 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human SOX4 protein | |
Breast or Colon Carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |