Learn More
BINDER™ Specimen Temperature Display
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB PRO Series
KB Series
KBF PRO Series
KBF Series
Unit Size:
Each
Description
Platinum 100 ohm temperature sensor for sample temperature display or control.
Specifications
Specifications
| For Use With (Equipment) | KB Series |
| Material | Steel |
| Type | Temperature Sensor |