Specifications
4 W | |
Shorwave UV tube w/ Longlife glass filter, white light flashlight | |
1.25 in. | |
8.24 in. |
254 nm | |
Mineralogy, stamp collecting, microbiology, mutation studies, laboratory UV dosing and illumination, and general white light inspection | |
3.17 cm |