Learn More
Spectroline™ MiniMax™ UV Lamp
Brand:  Spectroline™ UV-4FW
Encompass
 View more versions of this product
Product Code. 11510907
  • POA / Pack of 254
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Specifications
4 W
Shorwave UV tube w/ Longlife glass filter, white light flashlight
1.25 in.
8.24 in.
254 nm
Mineralogy, stamp collecting, microbiology, mutation studies, laboratory UV dosing and illumination, and general white light inspection
3.17 cm