Learn More
Spectroline™ MiniMax™ UV Lamp
Brand:  Spectroline™ UV-5F
Encompass
 View more versions of this product
Product Code. 11540907
  • POA / Pack of 254
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Specifications
5 W
Shorwave UV tube w/ Longlife glass filter
1.25 in.
9 in.
254 nm
Mineralogy, stamp collecting, microbiology, mutation studies, laboratory UV dosing and illumination
3.17 cm