Learn More
Spectroline™ Shortwave/Longwave UV Lamps
Brand:  Spectroline™ XX-15A
Encompass
 View more versions of this product
Product Code. 11550967
  • POA / Pack of 730
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Large, standard intensity units offer broad area, wide-angle coverage. At 10 in. (25cm), single-wavelength , double 15-watt tubes produce peak intensity of 1100μw/cm2 and combination wavelengths peak at 550μw/cm2 for longwave (UV-A), 490μw/cm2 for shortwave (UV-C). Shortwave units have Longlife™ transmitting filter, which ensures higher initial UV intensity and resistance to solarization. Overall dimensions: 18.75L × 4.5W × 3.5 in.H (47.6 × 11.4 × 8.9cm). For 115V 60Hz.
Order Info
All models feature rugged, anodized aluminum housing with mounting brackets.
Specifications
Specifications
Bench and Display UV Lamp
365 nm
3.5 in.
18.75 in.
1 Ea.
114.3 mm
UV-A
18.75 x 4.5 x 3.5 in. (476.2 x 114.3 x 88.9 mm)
88.9 mm
476.2 mm
4.5 in.