missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectroline™ UV Darkroom Cabinet
Portable, space-saving mini-cabinet doubles as a personal darkroom and UV irradiating units
Brand: Spectroline™ CM-26
This item is not returnable.
View return policy
Description
Portable, space-saving mini-cabinet doubles as a personal darkroom and UV irradiating units
- Flexible, contoured eyepiece with a built-in UV-absorbing window for safe sample viewing
- Felt curtain permits easy access to cabinet interior while shielding samples from external light
- Removable bottom panel allows easy positioning over transilluminators
- Lightweight, durable construction creates easily transportable darkroom
- Wavelength: UV (254nm-365nm)
- Wattage: 6w
- L×W×H: 30.5×16.5cm (12×6.5 in.)
Specifications
UV Darkroom Cabinet | |
6.5 in. | |
12 in. | |
1 Ea. | |
228.6 mm |
12 x 9 x 6.5 in. (304.8 x 228.6 x 165.1 mm) | |
165.1 mm | |
304.8 mm | |
9 in. |