Learn More
WTW™ Spectrophotometer photoLab™ 7100 VIS
GREENER_CHOICE
The spectral photometer photoLab™ 7100 VIS with AQA and IQ LabLink combines secured water analysis with uncomplicated special and spectral analytics. Provides the fastest and most affordable routine analysis via barcoded test kits for round and rectangular cuvettes. More than 250 methods are available for waste/drinking water, food and beverage industry as well as production, environmental monitoring or fish farming.
Brand:  WTW™ 250203
5142.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15335409
  • 5510.00 EUR / 1 unit
Estimated Shipment: 19-05-2025
Description
  • Easy to use: just place cuvette, read measurement value
  • More than 250 test programs for water analysis, galvanics and general lab analytics
  • Cell and reagent test kits with barcode for automatic program selection
  • Automatic cuvette and measurement range detection for rectangular cuvettes
  • Top reliability due to menu guided comprehensive Analytical Quality Assurance - AQA
  • Measurement Light on the road with car battery use
  • USB and Ethernet-connections for easy update, print to PDF or printer, storage and data export
  • photoLab™ Data spectral offers a GLP-compliant and convenient PC-interface to export, store and transfer data
  • IQ LabLink offers an outstanding connection between IQ Sensor Net and lab measurement for top measurement reliability
  • Measurement data of IQ Sensor Net sensors (VARiON) can be transferred via USB stick directly into photoLab 7100 VIS and back; faultless and without cable tangle thanks to sensor identification
  • Color determination by means of photometric measurement
  • With photoLab 7000/6000 Series and the PC-software, photoLab color common measurement procedures can be performed in a simple and neatly arranged way
Special procedures for environmental and food parameters such as chlorophyll can be executed simply and clearly arranged due to excellent menu guidance:
  • Programmed multistep- and multiwavelength methods
  • Comprehensive programming options for user defined requirements
  • Spectra and Kinetics measurements
  • Training of basics and modern photometry in education
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Spectrophotometer
Documents
Brochures
Product Title
WTW™ Spectrophotometer photoLab™ 7100 VIS >