Learn More
3M™ Speedglas™ Welding Helmet Front Cover
Click to view available options
:
Welding Helmet Cover
Description
Ensure you get the optimum level of comfort and protection from your welding helmet with 3M™ Welding Helmet Front Covers. Our spare parts replace worn or damaged front covers for the clear, protective visor of 3M™ Speedglas™ Welding Helmets. Make it a habit to check your equipment regularly for maximum safety.
Specifications
Specifications
| Certifications/Compliance | None |
| Color | Clear |
| For Use With (Equipment) | G5-02 Welding Helmets |
| Product Type | Front Panels |
| Size | Standard |