Learn More
Cole-Parmer™ Disposable Closed XRF X-Cell Sample Cups
Shop All SPEX Certiprep Products
Click to view available options
Diameter (English):
1.2 in.
1.57 in.
1.7 in.
Diameter (Metric):
31 mm
31 mm (micro)
40 mm
43 mm
Top Style:
Closed
Double Open Ended, Collar
Open, Collar
Unit Size:
Pack of 100
Description
Requires
Window film
- One opening for fast liquid sample delivery.
- Compatible with major suppliers of XRF spectrometers.
- Available in 31 mm, 40 mm, and 43 mm outer diameters.
- Contaminant-free polyethylene construction.
- Collars available to attach window film to cup for added security.
Cole-Parmer™ X-Cells are used throughout the world for fast, dependable, reproducible running of liquid samples in XRF spectrometers.
- They are designed to fit into the major suppliers of XRF spectrophotometers, such as PANalytical, Rigaku, Bruker, and Thermo Instruments.
- The durable polyethylene construction is generally resistant to acids, bases, and most organic chemicals except halogenated hydrocarbons.
- The patented closed X-Cell design features a snap-open seal for venting volatile samples and a reservoir cup for catching any liquids that expands through the vent.
- This assures the analyst of a smooth, fat window for improved precision, yet guards against spillage in inverted-optic spectrometers.
- A roughed surface within the reservoir cup facilitates labeling.
- Each closed X-Cell consists of a cup and snap-on ring.
- For routine liquid samples, the cup is filled with liquid, a piece of film is spready over the top, then the snap-ring is pushed down over the cup to form a fat drum-like window.
- The cell is turned over and, if desired, the snap-post on the rear can be pushed to break the seal and to permit venting of trapped bubbles and expand liquid while the sample is run.
- The filled X-Cell is then positioned in the spectrometer mask.
- Use the bubble-free insert with the 3527 (04576-05) 40 mm XRF liquid cell to prevent trapped air from forming a bubble under the window film when the cell is upright.
- The insert is pushed into the cell before the sample is added and forms an inverted-cone shelf under which air bubbles can be trapped once the cell is filled and sealed.
Order Info
Special application versions including the 31 mm micro X-Cell—with a 6.3 mm centered window for small, hazardous, or air-sensitive samples—and the 43 mm cell for Horiba™ sulfur analyzers also available. Supplied in bags of 100 cups.
Specifications
Specifications
| For Use With (Equipment) | XRF Spectrometers |
| Type | Sample Cup |
| Diameter (English) | 1.2 in. |
| Diameter (Metric) | 31 mm (micro) |
| For Use With (Application) | XRF Analysis |
| Material | Polyethylene |
| Quantity | 100 Pk. |
| Top Style | Open, Collar |