Learn More
Cytiva Spin Adapters for Gravity Columns
Shop All Cytiva Products
Click to view available options
Column Type:
For PD MidiTrap G-25 Column
For PD MiniTrap G-25 Column
For PD-10 Desalting Column
Unit Size:
Pack of 10
Description
- Convenient adapters, enable spinning of all gravity columns.
Specifications
Specifications
| Description | Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns. |
| Material | Polypropylene |
| Quantity | 10 Pack |
| For Use With (Application) | Convenient and rapid cleanup with high reproducibility for high throughput by running multiple samples in PD MidiTrap G-25 column for centrifugation |
| Type | Spin Column Adapter |
| Column Type | For PD MidiTrap G-25 Column |