missing translation for 'onlineSavingsMsg'
Learn More

SRP54 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

140.00 EUR - 399.00 EUR

Specifications

Antigen SRP54
Concentration 0.23 mg/mL
Applications Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16897994
View Documents Promotion Details
Proteintech
17976-1-AP-20UL
20 μL
140.00 EUR
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16887994
View Documents Promotion Details
Proteintech
17976-1-AP-150UL
150 μL
399.00 EUR
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody specifically recognizes the 54 kDa SRP54 protein.

Binds to the signal sequence of presecretory protein when they emerge from the ribosomes and transfers them to TRAM (translocating chain-associating membrane protein).
Specifications

Specifications

SRP54
Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Rat, Human
P14576, P61011, Q6AYB5
116650, 24067, 6729
SRP54 Fusion Protein Ag12166
Primary
-20°C
SRP54
0.23 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
SRP54
SRP54
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications