Learn More
Vitlab™ Stackable Graduated Beaker
Make your job easier and safer while handling a wide range of liquids with these polypropylene beakers. VITLAB™ Stackable Graduated Beakers feature an ergonomically designed handle and an easy to read, embossed black scale on both sides.
Brand:  Vitlab™ 482941
 View more versions of this product
Product Code. 10034690
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Highly transparent graduated beakers
  • Volume is equally visible for left- and right-handers
  • To preserve markings, cleaning at no higher than 60°C is recommended
Specifications
Specifications
Transparent
1000 mL
167 mm
Beaker
Yes
125 mm
Polypropylene
1 Ea.
Autoclavable
Black Printed Scale