Learn More
Vitlab™ Stackable Graduated Beaker
Make your job easier and safer while handling a wide range of liquids with these polypropylene beakers. VITLAB™ Stackable Graduated Beakers feature an ergonomically designed handle and an easy to read, embossed black scale on both sides.
Brand: Vitlab™ 481941
This item is not returnable.
View return policy
Description
- Highly transparent graduated beakers
- Volume is equally visible for left- and right-handers
- To preserve markings, cleaning at no higher than 60°C is recommended
Specifications
Transparent | |
500 mL | |
140 mm | |
Beaker | |
Yes |
100 mm | |
Polypropylene | |
1 Ea. | |
Autoclavable | |
Black Printed Scale |