Learn More
Bochem™ Stainless Steel Beaker
Click to view available options
Capacity (Metric):
100 mL
2000 mL
Unit Size:
Each
Description
- Stainless Steel
- For use with heating plates and magnetic stirrers
Specifications
Specifications
| Diameter (Metric) Base | 50mm |
| Capacity (Metric) | 100 mL |
| Material | Stainless Steel (18/10) |
| Height (Metric) | 60mm |
| Includes | With rim |