Learn More
Memmert™ Stainless Steel Bottom Drip Tray
Product Code. 17139664 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco, Not Option K1
HPP110eco, Not Option k1
HPP260eco
HPP410eco
HPP750eco, Not Option K1
HPP750eco, only with Option K1
IPP1060eco, Not Option K1
IPP110eco, Not Option K1
IPP260eco
IPP410eco
IPP750eco, Not Option K1
Unit Size:
Each
Product Code. 17139664
Brand: Memmert™ HPP110ecoB04359
Only null left
Add to basket
Add to basket
15 mm stainless steel bottom drip tray
  • May affect the temperature distribution
  • 15 mm
Specifications
Description Not in connection with option K1
For Use With (Equipment) HPP110eco, Not Option k1
Diameter (Metric) 15 mm