Learn More
Memmert™ Stainless Steel Bottom Drip Tray
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco, Not Option K1
HPP110eco, Not Option k1
HPP260eco
HPP410eco
HPP750eco, Not Option K1
HPP750eco, only with Option K1
IPP1060eco, Not Option K1
IPP110eco, Not Option K1
IPP260eco
IPP410eco
IPP750eco, Not Option K1
Unit Size:
Each
Description
- May affect the temperature distribution
- 15 mm
Specifications
Specifications
| Description | Not in connection with option K1 |
| For Use With (Equipment) | IPP110eco, Not Option K1 |
| Diameter (Metric) | 15 mm |