Learn More
AMSTA Rostfritt Stainless Steel Container with Lid
Product Code. p-200104597
Click to view available options
Capacity (Metric):
8 L
20 L
25 L
30 L
50 L
100 L
150 L
Includes:
Lid
Lid and 3 Clamps
Lid and 4 Clamps
Lid and 6 Clamps
Lid and Tap
Lid, Tap, and 3 Clamps
Lid, Tap, and 4 Clamps
Lid, Tap, and 6 Clamps
Unit Size:
Each
Product Code. 11741806
Brand: AMSTA Rostfritt 5060381
High-quality stainless steel container, available in various sizes with or without clamps and tap.
  • Quality
  • Invisible weld seams
  • Pharmaceutical grade finish
  • Sustainable production
  • Made in Sweden
Specifications
Product Type Can
Capacity (Metric) 8 L
Color Silver
Material Stainless Steel
Finish Ra 0.2 to 0.3 μm
Includes Lid
Height (Metric) 288 mm
Diameter (Metric) Outer 200 mm
Weight (Metric) 3.0 kg