Learn More
BINDER™ Expansion Rack
Expansion Rack constructed of stainless steel.
Brand: BINDER™ 8009-1092
This item is not returnable.
View return policy
Description
- Stainless steel
- Accessory rack
Specifications
Expansion racks - stainless steel | |
Rack | |
13.19 in. | |
15.12 in. | |
2.6 in. |
BINDER Chambers VD 56, VDL 56 | |
Stainless Steel | |
335 mm | |
384 mm | |
66 mm |