Learn More
BINDER™ Expansion Rack
Expansion Rack constructed of stainless steel.
Brand: BINDER™ 8009-1093
This item is not returnable.
View return policy
Description
- Stainless steel
- Accessory rack
Specifications
Expansion racks - stainless steel | |
Rack | |
11.14 in. | |
10.55 in. | |
2.2 in. |
BINDER Chambers VD 23, VDL 23 | |
Stainless Steel | |
283 mm | |
268 mm | |
56 mm |