missing translation for 'onlineSavingsMsg'
Learn More

GFL Gesellschaft Fuer Labortec™ Stainless Steel Fume Hoods for Waterbath

Use these baths for a varied range of applications. GFL Gesellschaft fuer Labortec™ Stainless Steel Fume Hoods for Waterbaths are ideal for gentle steaming work with Erlenmeyer flasks, glasses, etc.

Brand:  GFL Gesellschaft Fuer Labortec™ 1031

 View more versions of this product

Product Code. 10252601

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Bath, removable recessed lid with holes and perforated floor above the heating element are made of stainless steel
  • Adjustable water level regulator at the right-hand side of the unit is part of the standard scope of supply
  • Openings in the lid are covered by sets of rings made of heat-resistant plastic material
  • Temperature range from approx. 5°C above ambient to boiling point, temperature regulation through
  • tension thermostat
  • Four non-slip plastic feet ensure great stability
  • Heating element protected by over-temperature cut-out
Specifications

Specifications

5°C to 99.9°C
Waterbath
Ambient +5° to 99.9°C
Stainless Steel
300 mm
100 mm
91 mm
230 V
50/60 Hz
Tension Thermostat
Fume Hoods
Gentle steaming work with Erlenmeyer flasks, beakers, etc.
300 x 430 x 155 mm
155 mm
300 mm
5.7/7.0 kg
1.5 kW
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.