Learn More
Cole-Parmer™ Stainless Steel Funnel
Shop All Cole Parmer Products
Click to view available options
Capacity (Metric):
67 mL
70 mL
188 mL
350 mL
385 mL
658 mL
Height (Metric):
82.55 mm
104.1 mm
134.6 mm
160 mm
180.3 mm
210.8 mm
Unit Size:
Each
Description
Stainless steel funnels are a laboratory supply you will count on for years of use.
- Stay in stock with our selection of medium gauge, 304 stainless steel funnels
- Available in various capacities and overall sizes
Specifications
Specifications
| Capacity (Metric) | 188 mL |
| Material | Stainless Steel |
| Height (Metric) | 104.1 mm |