Learn More
Tenak™ Stainless Steel Sliding Shelf Rack
The Stainless Steel Sliding Shelf Rack is a comfort rack for upright freezers that allows a more ergonomic operating process when working with your samples. They are made of stainless steel and can be used with all freezer models.
42.05 EUR - 162.00 EUR
Specifications
Product Type | Sliding Shelf Rack |
---|---|
Holds | Boxes |
Material | Stainless Steel |
For Use With (Equipment) | Upright Freezers |
Quantity | 1 Ea. |
Product Code | Brand | Positions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Positions | Price | Quantity & Availability | |||||
12419550
|
Tenak™
TE24219 |
10 |
162.00 EUR
Each |
Estimated Shipment: 20-11-2024
Log in to see stock. |
|||||
15131289
|
Tenak™
TE25084 |
5 |
42.05 EUR
Each |
Estimated Shipment: 20-11-2024
Log in to see stock. |
|||||
Description
- Stainless steel
- Fit all Upright freezer models
Specifications
Sliding Shelf Rack | |
Stainless Steel | |
1 Ea. |
Boxes | |
Upright Freezers | |
140 mm |