Learn More
Nickel Electro™ Stainless Steel Gable Lid
Shop All Nickel-Electro Products
Click to view available options
For Use With (Equipment):
14 L Stirred Bath
22 L Stirred Bath
38 L Stirred Bath
8 L Stirred Bath
Unit Size:
Each
Description
- Stainless steel
- Insulated handle
- Minimises heat loss
- Reduces evaporation
- Assists in maintaining bath temperature
Specifications
Specifications
| Description | Stainless steel gable lid |
| For Use With (Equipment) | 8 L Stirred Bath |
| Material | Stainless Steel |
| Warranty | 3 Years |