missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Stainless Steel Stirring Paddles

Constructed of high quality stainless steel for durability. Sampling Systems™ Stainless Steel Stirring Paddles are ideal for use in Pharmaceutical and Hygienic applications.

Brand:  Sampling Systems™ A344-1000

 View more versions of this product

Product Code. 15829641

  • 122.00 EUR / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Crevice free construction
  • 316L Stainless Steel
  • Satin finish
  • Robust construction
  • BSE/TSE free
Specifications

Specifications

Paddle
Solid Blade
1000 mm
125 mm
Stainless Steel (316L)
Polished
240 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.