missing translation for 'onlineSavingsMsg'
Learn More

Standard Buffer Solution pH 4.01, SI Analytics™

Product Code. 11984719
Click to view available options
Quantity:
250 mL
60 Ampules
This item is not returnable. View return policy
This item is not returnable. View return policy

SI Analytics™ Standard Buffer Solution pH 4.01 resists changes in pH when small quantities of an acid are added to it.

  • Includes manufacturer certificate
  • Mixture, consisting of the following components: Water, potassium hydrogenphthalate
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Absorb with liquid-binding material (sand, diatomite, acid binders, universal binders, sawdust)
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
Color Colorless
Density 1.01g/cm3
pH 4.01
Physical Form Liquid
Quantity 250 mL
Formulation Water, potassium hydrogenphthalate
Odor Odorless
Packaging DURAN™ Bottle
Sterility Sterile
Vapor Pressure 23hPa
Solubility Information Fully miscible in water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.