missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Standard Float Switch
Install from the top of your tank, attach your own conduit to protect wiring
Brand: Cole-Parmer™ 07187-06
This item is not returnable.
View return policy
Description
- All models accept 120/240 VAC and operate as NO or NC switches except where noted
- Relay Output: at 24 VDC: 0.3 A; at 120 VAC: 0.17
- Min Secific Gravity: 0.9
- Switches include 24 in. leads (except 07187-12 which has 72 in. leads)
- If your level control system exceeds the current rating in the table, use a controller with your switch
Specifications
Polypropylene | |
24 in. Cable | |
-40°C |
2.09375 in. | |
150 psi |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction