Learn More
Macherey-Nagel™ Standard Silica 60 Adsorbents
Shop All Macherey-Nagel Products
Click to view available options
Type:
Standard Silica 60 Adsorbent
Unit Size:
25kg
Specifications
Specifications
| Particle Size | 0.04 to 0.063 mm |
| Pore Size | ∼60 Å |
| Technique | Column Chromatography |
| Quantity | 25 kg |
| Sorbent | Silica 60 |
| Surface Area (Metric) | ∼500 m2/g |
| Type | Standard Silica 60 Adsorbent |