Learn More
Brady™ Static Dissipative Thermal Transfer Printable Labels
Printable, static dissipative polyester labels for Thermal Transfer Printers.
77.95 EUR - 435.00 EUR
Specifications
Autoclavable | Not Autoclavable |
---|---|
Adhesive Type | Permanent Acrylic |
Color | White |
Finish | Gloss |
Material | Static-dissipative Polyester |
Product Code | Brand | Height (Metric) | Vertical Repeat (Metric) | Width (Metric) | Labels per Row | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Vertical Repeat (Metric) | Width (Metric) | Labels per Row | Price | Quantity & Availability | ||
11553664
|
Brady™
029909 |
5.1 mm | 7.6 mm | 12.7 mm | 5 |
141.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11578434
|
Brady™
054550 |
5.1 mm | 7.6 mm | 12.7 mm | 1 |
141.00 EUR
1 roll |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
11533664
|
Brady™
029907 |
5.1 mm | 7.6 mm | 16.5 mm | 4 |
157.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11598434
|
Brady™
054553 |
5.1 mm | 7.6 mm | 16.5 mm | 1 |
157.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11563654
|
Brady™
029900 |
6.4 mm | 8.9 mm | 19.1 mm | 4 |
157.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11518444
|
Brady™
054555 |
6.4 mm | 8.9 mm | 19.1 mm | 1 |
225.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11523674
|
Brady™
029916 |
6.4 mm | 8.9 mm | 22.9 mm | 1 |
212.00 EUR
1 roll |
Estimated Shipment: 02-01-2025
Log in to see stock. |
||
11593664
|
Brady™
029913 |
6.4 mm | 8.9 mm | 22.9 mm | 3 |
212.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11513674
|
Brady™
029915 |
6.4 mm | 8.9 mm | 31.8 mm | 1 |
237.00 EUR
1 roll |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
11583654
|
Brady™
029902 |
6.4 mm | 8.9 mm | 38.1 mm | 2 |
250.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11548444
|
Brady™
054558 |
6.4 mm | 8.9 mm | 38.1 mm | 1 |
261.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11533674
|
Brady™
029917 |
6.4 mm | 8.9 mm | 50.8 mm | 1 |
347.00 EUR
1 roll |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
11523664
|
Brady™
029906 |
6.4 mm | 9.5 mm | 13.8 mm | 2 |
237.00 EUR
1 roll |
Estimated Shipment: 02-01-2025
Log in to see stock. |
||
11503674
|
Brady™
029914 |
6.4 mm | 9.5 mm | 50.8 mm | 1 |
115.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11583664
|
Brady™
029912 |
6.4 mm | 8.9 mm | 6.4 mm | 7 |
162.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11503664
|
Brady™
029904 |
6.4 mm | 9.5 mm | 6.4 mm | 12 |
77.95 EUR
1 roll |
In Stock
Log in to see stock. |
||
11543664
|
Brady™
029908 |
7 mm | 9.5 mm | 12.7 mm | 4 |
167.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11573664
|
Brady™
029911 |
9.5 mm | 12.1 mm | 17.8 mm | 4 |
150.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11573654
|
Brady™
029901 |
9.5 mm | 12.1 mm | 25.4 mm | 3 |
237.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11528444
|
Brady™
054556 |
9.5 mm | 12.1 mm | 25.4 mm | 1 |
435.00 EUR
1 roll |
Estimated Shipment: 02-01-2025
Log in to see stock. |
||
11563664
|
Brady™
029910 |
9.5 mm | 12.1 mm | 9.5 mm | 7 |
170.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11513664
|
Brady™
029905 |
11.1 mm | 13.6 mm | 12.7 mm | 5 |
208.00 EUR
1 roll |
In Stock
Log in to see stock. |
||
11593654
|
Brady™
029903 |
12.7 mm | 15.2 mm | 25.4 mm | 3 |
339.00 EUR
1 roll |
Estimated Shipment: 02-01-2025
Log in to see stock. |
||
11538444
|
Brady™
054557 |
12.7 mm | 15.2 mm | 25.4 mm | 1 |
339.00 EUR
1 pack |
In Stock
Log in to see stock. |
||
11588434
|
Brady™
054552 |
12.7 mm | 15.2 mm | 12.7 mm | 1 |
224.00 EUR
1 pack |
In Stock
Log in to see stock. |
||
Description
- Material is UL and CSA approved with its respective ribbon
- Ideal for Circuit Board Labeling
- For smooth surfaces
Specifications
Not Autoclavable | |
White | |
Static-dissipative Polyester | |
Non-sterile | |
-40°C to +120°C |
Permanent Acrylic | |
Gloss | |
Thermal Transfer | |
BBP™81, BradyPrinter i5100, i7100, PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers | |
0.09 mm |