missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Staufen Rabbit anti-Human, Mouse, Rat, Clone: 8J5A6, Novus Biologicals™
Rabbit Monoclonal Antibody
196.00 EUR - 494.00 EUR
Specifications
Antigen | Staufen |
---|---|
Clone | 8J5A6 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18331646
|
Bio-Techne
NBP3-16450-20UL |
20 μg |
196.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18376966
|
Bio-Techne
NBP3-16450-100UL |
100 μg |
494.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Staufen Monoclonal antibody specifically detects Staufen in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Staufen | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6780 | |
IgG | |
Affinity purified |
8J5A6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
double-stranded RNA-binding protein Staufen homolog 1, FLJ25010, RNA binding protein (Drosophila), RNA-binding protein), staufen, RNA binding protein, homolog 1 (Drosophila) | |
A synthetic peptide corresponding to a sequence within amino acids 478-577 of human Staufen (O95793). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title