Learn More
Brady™ Maximum Chain Security
More secure than steel cables.
170.00 EUR - 266.00 EUR
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
15693537
|
Brady™
805846 |
Maximum Chain Security - 10 mm diam |
266.00 EUR
Pack of 2 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
11746925
|
Brady™
805846 |
Maximum Chain Security - 10 mm diam | N/A | |||||||
11736925
|
Brady™
805845 |
Maximum Chain Security - 8 mm diam | N/A | |||||||
15683537
|
Brady™
805845 |
Maximum Chain Security - 8 mm diam |
175.00 EUR
Pack of 2 |
In Stock
Log in to see stock. |
||||||
Description
- Supplied with a fabric sleeve which will protect paintwork and chrome from damage
Specifications
Chain | |
2 Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Brady™ Maximum Chain Security