Learn More
B Braun™ Sterican™ Intramuscular Cannula
Made of stainless chromium-nickel steel. B Braun™ Sterican™ Intramuscular Cannula is thin-walled with extremely smooth surface and light silicone coating. Special facet long grind provides pain-free punctures.
Brand: B Braun™ 4665473
This item is not returnable.
View return policy
Description
- According to ISO 7864 and DIN 13097
- Thin-walled cannula
- Made of stainless chromium-nickel steel
- With extremely smooth surface and light silicone coating
- Special facet long grind
- For pain-free punctures
- Transparent Luer-Lock plastic hub made of polypropylene
- Colour coded according to ISO 6009
- Pack of 100
Specifications
Stainless Chromium Nickel Steel | |
14 G | |
80 mm | |
1/Ea. |
Light Green | |
2.1 mm | |
Special Indication |