Learn More
BRAND™ Sterile Dispenser Cartridge
Sterile Dispenser Cartridge designed for use with Seripettor™, Seripettor™ Pro, and QuikSip.
Brand:  BRAND™ 704507
Product Code. 11688439
  • 110.00 EUR / Pack of 7
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Description
  • Constructed of PE/PP
  • For use with Seripettor™, Seripettor Pro™, and QuikSip
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyethylene, Polypropylene
Dispenser Cartridge
Seripettor™, Seripettor™ Pro, QuikSip
Sterile
2 mL