Learn More
Technical Service Consultants™ Sterile Inoculation Loop
Product Code. 12381309
Click to view available options
Volume:
1 μL
5 μL
10 μL
Color:
Clear
Red
White
Quantity:
1000 Pk.
20/bag
Unit Size:
Pack of 1000
Product Code. 12381309
Brand: Technical Service Consultants™ TS/4A
Only null left
Add to basket
Add to basket
Use the Technical Service Consultants™ Sterile Inoculation Loop for general purpose use or routine samples. Its soft HI polystyrene composition prevents the head breaking off, which makes it ideal for clinical use.
  • Laboratory use only
  • Shelf Life: 3 years.
  • Sterilization Method: Gamma Irradiation to achieve a SAL of 10-6
  • 4-25°C. Store in dry and ventilated area; protect from light, heat and moisture in tightly closed original container
Specifications
Disposable Single-use
Volume 1 μL
Material Polystyrene
Sterility Sterile
Color Clear
Product Type Inoculating Loop
For Use With (Equipment) Laboratory use only
Quantity 20/bag
For Use With (Application) Laboratory use only