Learn More
Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems
GREENER_CHOICE
Product Code. p-10155095 Shop All MBP Molecular BioProducts Products
Click to view available options
Volume (Metric):
30 μL
Unit Size:
Pack of 3840
Product Code. 13356048
Brand: Thermo Scientific™ 965261
Only null left
Add to basket
Add to basket
Increase performance through the superior tip straightness and low %CV with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems.
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 30 μL
Filtered Filtered
Sterility Sterile
Tip Style Extended Length
For Use With (Equipment) Beckman Coulter Liquid Handling Systems