Learn More
Thermo Scientific™ Sterilin™ Loops and Spreaders
Product Code. 11834732
Click to view available options
Volume:
1 μL
10 μL
Product Type:
Inoculating Loop and Needle, Flexible
Inoculating Loop and Needle, Rigid
Inoculating Loop, Spherical Tip
Inoculating Needle
L-Shaped Spreader
Color:
Blue
Dark blue
Dark green
Green
Pale Blue
Pale Green
Violet
Quantity:
10
500
1000
missing translation for 'unitSize'
Pack of 1000
Pack of 500
Pack of 800
Product Code. 11834732
missing translation for 'mfr': Thermo Scientific™ SL1S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Simplify your culture work with Thermo Scientific™ Sterilin™ Loops and Spreaders. Choices for inoculation, dilution streaking, spreading and colony picking.
Simplify your culture work with Thermo Scientific™ Sterilin™ Loops and Spreaders. Choices for inoculation, dilution streaking, spreading and colony picking.
NOT AVAILABLE IN AMERICA
  • For inoculation, dilution streaking, spreading and picking isolated colonies
  • Eliminate the need for flaming and the subsequent risk of aerosols Inoculation loop for fixed sample volumes with choice of sphere for dilution streaking, or needle for picking colonies and stab cultures
  • Choice of rigid or flexible loops to suit different applications and user preferences
  • Hexagonal shaft improves grip and assists orientation
  • Free of lubricants, oils and electrostatic charges to facilitate consistent wetting and complete liquid transfer
  • No rough edges means smooth, problem-free plating and streaking of cultures
  • Sterilized by gamma irradiation
Ordering Information
  • Not available in all geographic regions
TRUSTED_SUSTAINABILITY
Specifications
Volume 1 μL
Sterility Irradiated
Color Pale Green
Product Type Inoculating Loop and Needle, Flexible
Quantity 10