Learn More
Thermo Scientific™ Sterilin™ Weighing Boats
Click to view available options
Capacity (Metric):
5 mL
7 mL
30 mL
100 mL
250 mL
Color:
Black
White
Type:
Antistatic Weighing Dish
Standard
Weighing Boat
Shape:
Diamond
Square
Unit Size:
Pack of 500
Description
Form a funnel for easy pouring and complete sample transfer using the new and improved Thermo Scientific™ Sterilin™ Weighing Boats, featuring radiused corners. All sizes packed 250 per pack, 500 per case.
- Manufactured from high impact polystyrene
- Hydrophobic surface ensures no absorption of water from atmosphere or sample
- Available in black or white to ensure suitability for different colored sample materials
- Anti-static range alleviates issues with powder hang-up
- Flat base for a secure, firm rest on the balance pan
- Supplied nonsterile
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 7 mL |
| Color | Black |
| Material | Polystyrene |
| Shape | Square |
| Sterility | Non-sterile |
| Type | Antistatic Weighing Dish |
| Length (Metric) | 44 mm |
| Width (Metric) | 44 mm |