missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Sterisart Bd Bottled Media:Fluid A

Product Code. 15932710 Shop All Sartorius Products
Click to view available options
:
300 mL
This item is not returnable. View return policy

Product Code. 15932710

Brand: Sartorius Stedim Biotech™ BD254979

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sterility testing fluid A (peptone water) is used for diluting, dissolving, and rinsing of all samples.

  • Three different steps are required by the compendia depending on the condition of the sample:
    • Rinsing: Sterile devices or materials are rinsed in order to release adhered micro-organisms. After filtration of the sample, the Pharmacopoeia also require rinsing steps to be performed on the entire Sterisart NF unit.
    • Diluting: Small volumes or viscous liquids are diluted for better filterability.
    • Dissolving: Solid samples or ointments are dissolved for better filterability.
  • Fluid A (peptone water) is used for diluting, dissolving and rinsing of all types of samples.
  • Does not affect the viability of microorganisms
TRUSTED_SUSTAINABILITY

Specifications

Cap Type Type 2 Closure, 32 mm OD
Media Type Fluid A (Peptone Water)
Sterility Sterile
No. per Pack 10
Packaging Type Infusion Bottle
Quantity 300 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.