missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Sterisart Bd Bottled Media: FTM, Infusion Bottle
Shop All Sartorius Products
Click to view available options
:
100 mL
Description
- Fluid thioglycollate medium
- Liquid nutrient medium for the sterility testing of biologics and for the enrichment and cultivation of anaerobes, aerobes, and microaerophiles acc. to USP and EP.
- Not under vacuum, but the headspace of the FTM is gassed with a stream of nitrogen to maintain anaerobiosis.
- Contains a special quality of agar, resulting in a high clarity of the prepared medium..
Specifications
Specifications
| Cap Type | Type 2 Closure, 32 mm OD |
| Media Type | FTM (Fluid Thioglycollate Medium) |
| Sterility | Sterile |
| No. per Pack | 25 |
| Packaging Type | 125 mL Bottle |
| Quantity | 100 mL |
Safety and Handling
Product Identifier
- X10 Sterisart BD bottled Media, Type: FTM
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction