Learn More
Sampling Systems™ SteriWare Multi-grip Tweezers
Shop All Sampling Systems Products
Click to view available options
Length (Metric):
203 mm
300 mm
303 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 50
Description
- Made from food grade polystyrene
- Individually bagged to prevent cross contamination
- 100% manufactured and packed in a cleanroom
- Available in both sterile and non-sterile
Specifications
Specifications
| Tip Style | Serrated, Flat |
| Length (English) | 12 in. |
| Material | Polystyrene |
| Length (Metric) | 303 mm |
| Product Type | Tweezers |
| Quantity (No. per Case) | 50 Tweezers/Cs. |
| Sterility | Non-sterile |
| Style | Molded |