Learn More
Sampling Systems™ SteriWare Swabbing Templates
Shop All Sampling Systems Products
Click to view available options
Sterility:
Non-sterile
Sterile
Dimensions (L x W):
50 x 50 mm
100 x 100 mm
Unit Size:
Pack of 50
Description
- Available sterile or non-sterile
- Ensures consistent validation results
Specifications
Specifications
| Product Type | Swabbing Template |
| Material | HDPE |
| Autoclavable | Not Autoclavable |
| Disposable | Single-use |
| Graduated | No |
| Sterility | Sterile |
| Dimensions (L x W) | 50 x 50 mm |