Learn More
Fisherbrand™ Stainless Steel Stirring Shafts with Propellers
Product Code. 15143007 Shop All Fisherbrand Products
Click to view available options
Size:
305mm
457mm
610mm
762mm
Includes:
Propellor, 108mm
Propellor, 63.5mm
Unit Size:
Each
Product Code. 15143007
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Excellent at producing axial flows in vessels from top to bottom. Fisherbrand™ Stainless Steel Stirring Shafts with Propellers are for use with overhead stirrer units.
  • Available in a range of sizes
  • Resistant to corrosion
Specifications
Includes Propellor, 108mm
Quantity 1 Each
Size 457mm
Type Stirring Shaft