missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Streptavidin, Alexa Fluor™ 610—R-Phycoerythrin Conjugate (Alexa Fluor™ 610—R-Phycoerythrin Streptavidin)

Streptavidin, Alexa Fluor™ 610—R-Phycoerythrin Conjugate (Alexa Fluor™ 610—R-Phycoerythrin Streptavidin)

Brand:  Molecular Probes™ S20982

Product Code. 10277522

  • 634.00 EUR / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The streptavidin, Alexa Fluor™ 610-R-phycoerythrin streptavidin tandem conjugate has excitation and emission maxima at ∼496,565/627 and is widely used to detect biotinylated probes by flow cytometry. Not only are the Alexa Fluor™ 610 conjugates of R-PE are more fluorescent than the commercially available Texas Red™ conjugates of R-PE, the fluorescent emission of Alexa Fluor™ 610-R-PE tandem conjugates is shifted to somewhat longer wavelengths than is the emission of Texas Red™-R-PE conjjgates, resulting in better separation from the emission of R-PE.

View a selection guide for all Alexa Fluor™ streptavidin and avidin conjugates.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mg/mL
Fluorescence
100 μL
488 or 561/630
Wet Ice
Alexa Fluor™
PE-Alexa Fluor 610
PE-Alexa Fluor 610
Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Contains 1 vial of streptavidin R-PE-Alexa Fluor™ 610 conjugate (1 mg/mL).

Store in refrigerator (2–8°C) and protect from light.
488 or 561
Solution
Streptavidin Conjugate (Fluorescent)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.