missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Streptavidin, BV421
Click to view available options
Quantity:
100 μg
Unit Size:
Each
Description
Species Reactivity:Ms
Flow Cytometry
Specifications
Specifications
| Concentration | 0.1 mg/mL |
| Quantity | 100 μg |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction