Learn More
BD Streptavidin, PE-CF594
Developed as a better alternative to PE-Texas Red, for use in Flow Cytometry
234.00 EUR - 619.00 EUR
Specifications
Concentration | 0.2 mg/mL |
---|---|
Conjugate | PE-CF594 |
Content And Storage | 4°C |
For Use With (Application) | Flow Cytometry |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15801819
|
BD
562318 |
25 μg |
234.00 EUR
Each |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
15861779
|
BD
562284 |
0.1 mg |
619.00 EUR
Each |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
Description
The Streptavidin is conjugated to BD Horizon PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496 nm and 564 nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red (eg 610/20-nm filter).
Specifications
0.2 mg/mL | |
PE-CF594 | |
4°C | |
Flow Cytometry |